(2-Hydroxyethyl)triphenylphosphonium bromide 96%

Supplier: Ambeed
Warning

A174574-100G A174574-25G A174574-5G
77138-518EA 148.53 USD
77138-518 77138-520 77138-522
(2-Hydroxyethyl)triphenylphosphonium bromide 96%
(2-Hydroxyethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₀H₂₀BrOP
MW: 387.26 g/mol
MDL Number: MFCD00011901
CAS Number: 7237-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR