(Diethoxy(methyl)silyl)methyl methacrylate 97%
Supplier: Ambeed
|
Warning
|
A174270-25G
A174270-5G
77138-512EA
61.8
USD
77138-512
77138-514
(Diethoxy(methyl)silyl)methyl methacrylate 97%
(Diethoxy(methyl)silyl)methyl methacrylate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀O₄Si |
MDL Number:
MFCD00545072 CAS Number: 3978-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...