(R)-3-Amino-3-(o-tolyl)propanoic acid 95%

Supplier: Ambeed
Warning

A173669-100MG A173669-250MG
77138-486EA 36.08 USD
77138-486 77138-488
(R)-3-Amino-3-(o-tolyl)propanoic acid 95%
(R)-3-Amino-3-(o-tolyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD04113646
CAS Number: 752198-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR