Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: RuPhos-Pd-G3

A172748-10G A172748-250MG A172748-5G A172748-1G A172748-25G
77138-460EA 456.19 USD
77138-460 77138-462 77138-464 77550-112 77567-542
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: For research use only.

Formula: C₄₃H₅₆NO₅PPdS
MW: 836.37 g/mol
MDL Number: MFCD22572684
CAS Number: 1445085-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR