(5-Chloro-2-iodophenyl)(2,6-difluorophenyl)methanone 98%

Supplier: Ambeed
Warning

A172533-100MG A172533-1G
77138-456EA 56.64 USD
77138-456 77138-458
(5-Chloro-2-iodophenyl)(2,6-difluorophenyl)methanone 98%
(5-Chloro-2-iodophenyl)(2,6-difluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₆ClF₂IO
MDL Number: MFCD10565698
CAS Number: 869365-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR