2-Chloro-4-(trifluoromethoxy)aniline 97%

Supplier: Ambeed
Warning

A172378-10G A172378-25G A172378-5G A172378-1G
77138-450EA 29.87 USD
77138-450 77138-452 77138-454 77685-384
2-Chloro-4-(trifluoromethoxy)aniline 97%
2-Chloro-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MDL Number: MFCD01631704
CAS Number: 69695-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR