2-Bromo-4-chloroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-chloro-1-methoxybenzene

A171317-100G A171317-10G A171317-25G
77138-424EA 47.75 USD
77138-424 77138-426 77138-428
2-Bromo-4-chloroanisole 97%
2-Bromo-4-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD00079705
CAS Number: 60633-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR