D-(-)-Diisopropyl tartrate 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester

A171104-10G A171104-25G
77138-402EA 10.61 USD
77138-402 77138-404
D-(-)-Diisopropyl tartrate 98%
D(-)-Diisopropyl tartrate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
MDL Number: MFCD00008876
CAS Number: 62961-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR