1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 4'-Amino-2,2,2-trifluoroacetophenone, 1-(4-Aminophenyl)-2,2,2-trifluoroethanone

A170552-100MG A170552-1G A170552-250MG A170552-5G
77138-394EA 27.59 USD
77138-394 77138-396 77138-398 77138-400
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone 95%
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
MDL Number: MFCD05663790
CAS Number: 23516-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR