2-(2-Chloro-5-nitrophenyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
A169597-100MG
A169597-1G
A169597-250MG
77138-386EA
30.77
USD
77138-386
77138-388
77138-390
2-(2-Chloro-5-nitrophenyl)pyridine 97%
2-(2-Chloro-5-nitrophenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₇ClN₂O₂ MW: 234.64 g/mol |
MDL Number:
MFCD12024454 CAS Number: 879088-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...