(6-Isopropyl-9-methyl-1,4-dioxaspiro[4.5]decan-2-yl)methanol 98%

Supplier: Ambeed
Danger

A169251-5G A169251-25G
77138-374EA 11.67 USD
77138-374 77615-406
(6-Isopropyl-9-methyl-1,4-dioxaspiro[4.5]decan-2-yl)methanol 98%
(6-Isopropyl-9-methyl-1,4-dioxaspiro[4.5]decan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₂₄O₃
MW: 228.32 g/mol
MDL Number: MFCD18083746
CAS Number: 63187-91-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR