(-)-Corey lactone benzoate 97%

Supplier: Ambeed
Warning

Synonyms: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Benzoate, (3aR,4S,5R,6aS)-5-(Benzoyloxy)hexahydro-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

A169100-100MG A169100-1G A169100-250MG A169100-25G
77138-366EA 7.43 USD
77138-366 77138-368 77138-370 77685-369
(-)-Corey lactone benzoate 97%
(-)-Corey lactone benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₅
MW: 276.29 g/mol
MDL Number: MFCD00674102
CAS Number: 39746-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR