(S)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride 98%

Supplier: Ambeed
Warning

A168437-1G A168437-250MG A168437-5G A168437-100MG
77138-350EA 180.25 USD
77138-350 77138-352 77138-354 77685-365
(S)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride 98%
(S)-N-Methyl-N-(pyrrolidin-3-yl)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂O
MDL Number: MFCD29918571
CAS Number: 1215264-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR