2-Bromo-6-chloronicotinonitrile 97%

Supplier: Ambeed
Warning

A167328-100MG A167328-1G A167328-250MG A167328-50MG
77138-342EA 80.63 USD
77138-342 77138-344 77138-346 77138-348
2-Bromo-6-chloronicotinonitrile 97%
2-Bromo-6-chloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.45 g/mol
MDL Number: MFCD18257840
CAS Number: 1256833-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR