3-Hydroxy-1-adamantaneacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Carboxymethyl)-3-hydroxyadamantane, 2-(3-Hydroxyadamantan-1-yl)acetic acid

A166681-1G A166681-250MG A166681-25G A166681-5G
77138-324EA 19.1 USD
77138-324 77138-326 77138-328 77138-330
3-Hydroxy-1-adamantaneacetic acid 97%
3-Hydroxy-1-adamantaneacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₃
MW: 210.27 g/mol
MDL Number: MFCD00167797
CAS Number: 17768-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR