4-Chloro-3-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

A165734-100G A165734-25G A165734-5G
77138-300EA 126.68 USD
77138-300 77138-302 77138-304
4-Chloro-3-nitrobenzyl alcohol 98%
4-Chloro-3-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD00007086
CAS Number: 55912-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR