(3-Mercaptopropyl)trimethoxysilane 98%

Supplier: Ambeed
Danger

Synonyms: Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol

A165640-25G A165640-100G
77138-298EA 6.37 USD
77138-298 77593-000
(3-Mercaptopropyl)trimethoxysilane 98%
(3-Mercaptopropyl)trimethoxysilane

Precaución: for Research Use Only

Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
MW: 196.34 g/mol
MDL Number: MFCD00004901
CAS Number: 4420-74-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR