MHY1485 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dimorpholino-N-(4-nitrophenyl)-1,3,5-triazin-2-amine

A165303-10MG A165303-1MG A165303-250MG A165303-5MG A165303-100MG A165303-50MG
77138-288EA 41.38 USD
77138-288 77138-290 77138-292 77138-294 77685-344 77685-345
MHY1485 98%
MHY1485

Precaución: for Research Use Only

Formula: C₁₇H₂₁N₇O₄
MW: 387.39 g/mol
MDL Number: MFCD00489974
CAS Number: 326914-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR