(3-Bromopropyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromopropyltriphenylphosphonium Bromide

A163726-100G A163726-10G A163726-500G A163726-25G
77138-246EA 23.68 USD
77138-246 77138-248 77138-250 77320-130
(3-Bromopropyl)triphenylphosphonium bromide 97%
(3-Bromopropyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₁Br₂P
MW: 464.18 g/mol
MDL Number: MFCD00011866
CAS Number: 3607-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR