4'-Demethylepipodophyllotoxin 98%

Supplier: Ambeed
Warning

Synonyms: (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one

A163397-50MG A163397-1G A163397-5G A163397-25G
77138-244EA 5.31 USD
77138-244 77567-386 77615-344 77685-336
4'-Demethylepipodophyllotoxin 98%
4'-Demethylepipodophyllotoxin

Precaución: For research use only.

Formula: C₂₁H₂₀O₈
MW: 400.38 g/mol
MDL Number: MFCD00189421
CAS Number: 6559-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR