5,6-Dibromopyridin-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2,3-dibromopyridine

A163087-100MG A163087-10G A163087-1G A163087-250MG
77138-236EA 37.14 USD
77138-236 77138-238 77138-240 77138-242
5,6-Dibromopyridin-3-amine 95%
5,6-Dibromopyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD11110698
CAS Number: 53242-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR