Darunavir ethanolate 98+%

Supplier: Ambeed
Warning

Synonyms: (3R,3AS,6aR)-hexahydrofuro[2,3-b]furan-3-yl ((2S,3R)-4-(4-amino-N-isobutylphenylsulphonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate compound with ethanol (1:1), (3R,3AS,6aR)-Hexahydrofuro[2,3-b]furan-3-yl ((2S,3R)-4-(4-amino-N-isobutylphenylsulphonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate Ethanolate

A162616-10MG A162616-1MG A162616-50MG A162616-5G A162616-5MG
77138-206EA 9.55 USD
77138-206 77138-208 77138-210 77138-212 77138-214
Darunavir ethanolate 98+%
Darunavir ethanolate

Precaución: for Research Use Only

Formula: C₂₉H₄₃N₃O₈S
MW: 593.736 g/mol
MDL Number: MFCD18251642
CAS Number: 635728-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR