1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

A161372-1G A161372-250MG
77138-178EA 87 USD
77138-178 77138-180
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester 97%
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₃
MW: 264.13 g/mol
MDL Number: MFCD09037503
CAS Number: 1072944-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR