(2-Acetoxyethoxy)methyl Acetate 97%

Supplier: Ambeed
Warning

A160660-100G A160660-10G A160660-25G A160660-500G A160660-5G A160660-1000G
77138-138EA 16.48 USD
77138-138 77138-140 77138-142 77138-144 77138-146 77685-317
(2-Acetoxyethoxy)methyl Acetate 97%
(2-Acetoxyethoxy)methyl Acetate

Precaución: for Research Use Only

Formula: C₇H₁₂O₅
MDL Number: MFCD09031327
CAS Number: 59278-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR