(3-Chloro-2-hydroxypropyl)trimethylammonium chloride 65% in water

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride(65% in water), 3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride

A160561-1000G A160561-100G A160561-25G A160561-500G
77138-118EA 55.17 USD
77138-118 77138-120 77138-122 77138-124
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride 65% in water
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride

Precaución: for Research Use Only

Formula: C₆H₁₅Cl₂NO
MW: 188.09 g/mol
MDL Number: MFCD00055655
CAS Number: 3327-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR