Nintedanib ethanesulfonate salt 98%

Supplier: Ambeed
Warning

Synonyms: (Z)-Methyl 3-(((4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxylate ethanesulphonate, Nintedanib esylate

A160216-50MG A160216-10MG A160216-1G A160216-250MG A160216-5G
77138-112EA 5.31 USD
77138-112 77685-308 77685-309 77685-310 77685-311
Nintedanib ethanesulfonate salt 98%
Nintedanib ethanesulfonate salt

Precaución: for Research Use Only

Formula: C₃₃H₃₉N₅O₇S
MW: 649.76 g/mol
MDL Number: MFCD26142360
CAS Number: 656247-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR