(S)-(+)-Carvone 98%

Supplier: Ambeed
Warning

A160210-100G A160210-25G A160210-500G A160210-5G
77138-104EA 55.36 USD
77138-104 77138-106 77138-108 77138-110
(S)-(+)-Carvone 98%
(S)-(+)-Carvone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00062997
CAS Number: 2244-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR