(R)-4-Methoxy-2-methyl-4-oxobutanoic acid 95%
Supplier: Ambeed
|
Warning
|
A160192-100MG
A160192-1G
A160192-250MG
77138-098EA
105.03
USD
77138-098
77138-100
77138-102
(R)-4-Methoxy-2-methyl-4-oxobutanoic acid 95%
(R)-4-Methoxy-2-methyl-4-oxobutanoic acid
Precaución: for Research Use Only
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol |
MDL Number:
MFCD00211259 CAS Number: 81025-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...