Tetrabutylphosphonium tetrafluoroborate 98%

Supplier: Ambeed
Danger

A159900-1G A159900-25G A159900-5G
77138-088EA 7.21 USD
77138-088 77138-090 77138-092
Tetrabutylphosphonium tetrafluoroborate 98%
Tetrabutylphosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₆H₃₆BF₄P
MDL Number: MFCD00270144
CAS Number: 1813-60-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR