4-Nitro-2-(trifluoromethyl)aniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine
A159502-100G
A159502-10G
A159502-25G
A159502-5G
77138-070EA
33.99
USD
77138-070
77138-072
77138-074
77138-076
77138-078
4-Nitro-2-(trifluoromethyl)aniline 98%
4-Nitro-2-(trifluoromethyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₅F₃N₂O₂ MW: 206.12 g/mol |
MDL Number:
MFCD00007365 CAS Number: 121-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...