Propargyl-PEG1-acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Prop-2-yn-1-yloxy)propanoic acid

A158981-100MG A158981-5G A158981-1G A158981-250MG
77138-054EA 20.6 USD
77138-054 77138-056 77549-998 77685-304
Propargyl-PEG1-acid 95%
Propargyl-PEG1-acid

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.126 g/mol
MDL Number: MFCD10689189
CAS Number: 55683-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR