3-Amino-2,6-difluorobenzoic acid 95%

Supplier: Ambeed
Warning

A158559-1G A158559-5G
77138-050EA 16.47 USD
77138-050 77138-052 77685-303
3-Amino-2,6-difluorobenzoic acid 95%
3-Amino-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C7H5F2NO2
MDL Number: MFCD07368833
CAS Number: 83141-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR