2-Hydroxy-3,5-dibromo-4-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-2-hydroxy-4-methylpyridine

A158230-100MG A158230-10G A158230-1G A158230-250MG A158230-5G
77138-040EA 23.34 USD
77138-040 77138-042 77138-044 77138-046 77138-048
2-Hydroxy-3,5-dibromo-4-methylpyridine 95%
2-Hydroxy-3,5-dibromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
MDL Number: MFCD06254604
CAS Number: 89581-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR