Methyl 6-hydroxy-5-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

A158096-100MG A158096-1G A158096-250MG
77138-034EA 126.25 USD
77138-034 77138-036 77138-038
Methyl 6-hydroxy-5-(trifluoromethyl)picolinate 97%
Methyl 6-hydroxy-5-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MW: 221.13 g/mol
MDL Number: MFCD28039874
CAS Number: 1006364-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR