1,2-Dibromo-3,4-difluorobenzene 95%

Supplier: Ambeed
Warning

A157895-100MG A157895-250MG
77138-030EA 23.69 USD
77138-030 77138-032
1,2-Dibromo-3,4-difluorobenzene 95%
1,2-Dibromo-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MDL Number: MFCD12922587
CAS Number: 811713-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR