2-Bromo-5-chlorotoluene 97%
Supplier: Ambeed
|
Warning
|
A157714-1000G
A157714-100G
A157714-10G
A157714-25G
A157714-500G
A157714-5G
77138-018EA
244.11
USD
77138-018
77138-020
77138-022
77138-024
77138-026
77138-028
2-Bromo-5-chlorotoluene 97%
2-Bromo-5-chlorotoluene
Precaución: for Research Use Only
Formula:
C₇H₆BrCl |
MDL Number:
MFCD00000602 CAS Number: 14495-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...