4-Bromo-2-nitrobenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-nitrobenzylalcohol

A157316-10G A157316-1G A157316-250MG A157316-5G
77138-006EA 115.64 USD
77138-006 77138-008 77138-010 77138-012
4-Bromo-2-nitrobenzyl alcohol 95%
4-Bromo-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.031 g/mol
MDL Number: MFCD07779493
CAS Number: 22996-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR