Pyridine-2,6-dicarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA
A156557-1000G
A156557-100G
A156557-10G
A156557-25G
A156557-500G
77137-966EA
182.32
USD
77137-966
77137-968
77137-970
77137-972
77137-974
Pyridine-2,6-dicarboxylic acid 98%
Pyridine-2,6-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅NO₄ MW: 167.12 g/mol Boiling Pt: 357 Melting Pt: 327 Flash Pt: 188 |
MDL Number:
MFCD00006299 CAS Number: 499-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...