(R)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid 97%

Supplier: Ambeed
Warning

A156469-100MG A156469-10G A156469-1G A156469-250MG A156469-5G
77137-950EA 6.37 USD
77137-950 77137-952 77137-954 77137-956 77137-958
(R)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid 97%
(R)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₄S
MW: 319.38 g/mol
MDL Number: MFCD15071654
CAS Number: 86117-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR