Selonsertib 97%

Supplier: Ambeed
Warning

Synonyms: GS-4997, 5-(4-cyclopropyl-1H-imidazol-1-yl)-2-fluoro-N-(6-(4-isopropyl-4H-1,2,4-triazol-3-yl)pyridin-2-yl)-4-methylbenzamide

A156190-100MG A156190-250MG A156190-1MG A156190-5MG
77137-946EA 107.16 USD
77137-946 77137-948 77685-289 77685-290
Selonsertib 97%
Selonsertib

Precaución: for Research Use Only

Formula: C₂₄H₂₄FN₇O
MW: 445.5 g/mol
MDL Number: MFCD29920355
CAS Number: 1448428-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR