(6-(Cyclohexyloxy)-2-fluoropyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A155740-1G A155740-250MG A155740-5G
77137-928EA 156.57 USD
77137-928 77137-930 77137-932
(6-(Cyclohexyloxy)-2-fluoropyridin-3-yl)boronic acid 97%
(6-(Cyclohexyloxy)-2-fluoropyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MDL Number: MFCD18434426
CAS Number: 2096332-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR