(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-Ph-BPE, 1,2-Bis((2R,5R)-2,5-diphenylphospholan-1-yl)ethane

A155397-100MG A155397-250MG A155397-50MG A155397-1G A155397-5G
77137-922EA 155.96 USD
77137-922 77137-924 77137-926 77549-974 77685-283
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane 98%
(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane

Precaución: for Research Use Only

Formula: C₃₄H₃₆P₂
MW: 506.61 g/mol
MDL Number: MFCD07369027
CAS Number: 528565-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR