(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-1,1,2-Triphenyl-1,2-ethanediol

A155244-1G A155244-250MG A155244-5G
77137-916EA 22.66 USD
77137-916 77137-918 77137-920
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol 98%
(R)-(+)-1,1,2-Triphenyl-1,2-ethanediol

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₂
MDL Number: MFCD00134424
CAS Number: 95061-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR