(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone 98%

Supplier: Ambeed
Warning

A154920-10G A154920-1G A154920-5G A154920-100G
77137-910EA 24.73 USD
77137-910 77137-912 77137-914 77746-062
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone 98%
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MDL Number: MFCD04973958
CAS Number: 40856-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR