1-Ethylpropylamine 98%

Supplier: Ambeed
Danger

A154678-10G A154678-25G
77137-900EA 16.48 USD
77137-900 77567-252
1-Ethylpropylamine 98%
1-Ethylpropylamine

Precaución: For research use only.

Formula: C₅H₁₃N
MDL Number: MFCD00008096
CAS Number: 616-24-0
UN: 2733
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR