Vidarabine 98%

Supplier: Ambeed
Warning

Synonyms: Adenine 9-β-D-arabinofuranoside, 9-β-D-Arabinofuranosyladenine, (2R,3S,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-β-D-Arabinofuranosyladenine, Ara-A, Vidarabine

A153495-10G A153495-5G A153495-25G A153495-100G A153495-1G A153495-500G
77137-884EA 14.86 USD
77137-884 77137-886 77549-942 77567-230 77600-182 77615-250
Vidarabine 98%
Vidarabine

Precaución: For research use only.

Formula: C₁₀H₁₃N₅O₄
MW: 267.24 g/mol
MDL Number: MFCD00065471
CAS Number: 5536-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR