4-Chloro-6-nitro-o-toluidine 98%

Supplier: Ambeed
Warning

A153126-1G A153126-25G A153126-5G
77137-878EA 17.51 USD
77137-878 77137-880 77137-882
4-Chloro-6-nitro-o-toluidine 98%
4-Chloro-6-nitro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MDL Number: MFCD01320687
CAS Number: 62790-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR