4-Chloro-3-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(4-chloro-3-(trifluoromethyl)phenyl)borane, 4-Chloro-3-(trifluoromethyl)benzeneboronic acid, (4-Chloro-3-(trifluoromethyl)phenyl)metaboric acid

A152882-10G A152882-1G A152882-25G A152882-5G
77137-846EA 58.35 USD
77137-846 77137-848 77137-850 77137-852
4-Chloro-3-(trifluoromethyl)phenylboronic acid 97%
4-Chloro-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD03094999
CAS Number: 176976-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR