4,6-Dimethoxy-5-methylpyrimidine 97%

Supplier: Ambeed
Warning

A152320-100MG A152320-1G
77137-822EA 30.77 USD
77137-822 77137-824
4,6-Dimethoxy-5-methylpyrimidine 97%
4,6-Dimethoxy-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
MDL Number: MFCD00196466
CAS Number: 13566-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR