(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)non-8-enoic acid 97%

Supplier: Ambeed
Warning

A152032-100MG A152032-250MG
77137-806EA 108.22 USD
77137-806 77137-808
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)non-8-enoic acid 97%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)non-8-enoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₄
MW: 393.48 g/mol
MDL Number: MFCD19381837
CAS Number: 1262886-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR